missing translation for 'onlineSavingsMsg'
Learn More

Cathepsin B Antibody (004), DyLight 755, Novus Biologicals™

Product Code. 18394171 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18394171 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18394171 Supplier Novus Biologicals Supplier No. NBP290401IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Cathepsin B Monoclonal specifically detects Cathepsin B in Mouse samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cathepsin B
Applications ELISA
Classification Monoclonal
Clone 4
Conjugate DyLight 755
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias APP secretase, APPS, cathepsin B, Cathepsin B1, CPSBamyloid precursor protein secretase, cysteine protease, EC 3.4.22, EC 3.4.22.1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Cathepsin B (P10605; Met1-Phe339).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Cancer, Cellular Markers, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1508
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.