missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15020-1-AP-150UL
This item is not returnable.
View return policy
Description
The cathepsin family of proteolytic enzymes include several diverse classes of proteases. Cathepsins B, L, H, K, S and O comprise the cysteine protease class. Cathepsins D and E comprise the aspartyle protease class. The serine protease class includes cathepsin G. Cathepsins function in cellular metabolism and participate in peptide biosynthesis and protein degradation. Cathepsin A, a serine carboxypeptidase, exists in a high molecular weight lysosomal complex with b-galactosidase (b-gal) and a-neuraminidase (Neu1). Cathepsin A functions to protect b-gal and Neu1 from intralysosomal proteolysis. Deficiencies in cathepsin A lead to deficiencies in b-gal and Neu1. The gene encoding human cathepsin A maps to chromosome 20q13.12. Mutations in this gene cause glactosialidosis, a lysosomal storage disorder resulting from the b-gal and Neu1 deficiencies.Specifications
| Cathepsin A | |
| Polyclonal | |
| Unconjugated | |
| CTSA | |
| Carboxypeptidase C, Carboxypeptidase L, Cathepsin A, CTSA, GLB2, GSL, Lysosomal protective protein, NGBE, PPCA, PPGB, Protective protein cathepsin A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19025, 296370, 5476 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.46 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P10619, P16675 | |
| Ctsa | |
| Cathepsin A Fusion Protein Ag7073 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction