missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cathepsin A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102793
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CTSA.
The cathepsin family of proteolytic enzymes include several diverse classes of proteases. Cathepsins B, L, H, K, S and O comprise the cysteine protease class. Cathepsins D and E comprise the aspartyle protease class. The serine protease class includes cathepsin G. Cathepsins function in cellular metabolism and participate in peptide biosynthesis and protein degradation. Cathepsin A, a serine carboxypeptidase, exists in a high molecular weight lysosomal complex with b-galactosidase (b-gal) and a-neuraminidase (Neu1). Cathepsin A functions to protect b-gal and Neu1 from intralysosomal proteolysis. Deficiencies in cathepsin A lead to deficiencies in b-gal and Neu1. The gene encoding human cathepsin A maps to chromosome 20q13.12. Mutations in this gene cause glactosialidosis, a lysosomal storage disorder resulting from the b-gal and Neu1 deficiencies.
Specifications
| Cathepsin A | |
| Polyclonal | |
| Unconjugated | |
| CTSA | |
| AU019505; beta-galactosidase 2; beta-galactosidase protective protein; carboxypeptidase C; Carboxypeptidase L; carboxypeptidase Y-like kininase; carboxypeptidase-L; cathepsin A; CTSA; deamidase; GLB2; GSL; lysosomal carboxypeptidase A; lysosomal protective protein; Lysosomal protective protein 20 kDa chain; Lysosomal protective protein 32 kDa chain; NGBE; PPCA; PPGB; protective protein cathepsin A; Protective protein for beta-galactosidase; urinary kininase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 19025, 296370, 5476 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P10619, P16675 | |
| CTSA | |
| A synthesized peptide derived from human CTSA(Accession P10619), corresponding to amino acid residues L394-F444. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction