missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin A/Lysosomal Carboxypeptidase A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98264-50ul
This item is not returnable.
View return policy
Description
Cathepsin A/Lysosomal Carboxypeptidase A Polyclonal antibody specifically detects Cathepsin A/Lysosomal Carboxypeptidase A in Mouse samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| Cathepsin A/Lysosomal Carboxypeptidase A | |
| Polyclonal | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 4-6 μL/mg of lysate | |
| beta-galactosidase 2, beta-galactosidase protective protein, Carboxypeptidase C, Carboxypeptidase L, cathepsin ANGBE, EC 3.4.16, EC 3.4.16.5, GLB2, GSL, lysosomal protective protein, PPCA, PPGBprotective protein for beta-galactosidase (galactosialidosis), Protective protein cathepsin A, Protective protein for beta-galactosidase | |
| Produced in rabbits immunized with purified, recombinant Mouse Cathepsin A/Lysosomal Carboxypeptidase A (Uniprot#: P16675-1; Met1-Tyr474) | |
| 50 μg | |
| Signal Transduction | |
| 5476 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction