missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin A/Lysosomal Carboxypeptidase A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98190-50ul
This item is not returnable.
View return policy
Description
Cathepsin A/Lysosomal Carboxypeptidase A Polyclonal antibody specifically detects Cathepsin A/Lysosomal Carboxypeptidase A in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Cathepsin A/Lysosomal Carboxypeptidase A | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:50-1:200 | |
| beta-galactosidase 2, beta-galactosidase protective protein, Carboxypeptidase C, Carboxypeptidase L, cathepsin ANGBE, EC 3.4.16, EC 3.4.16.5, GLB2, GSL, lysosomal protective protein, PPCA, PPGBprotective protein for beta-galactosidase (galactosialidosis), Protective protein cathepsin A, Protective protein for beta-galactosidase | |
| Produced in rabbits immunized with E. coli-derived Human Cathepsin A/Lysosomal Carboxypeptidase A fragment. | |
| 50 μg | |
| Signal Transduction | |
| 5476 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction