missing translation for 'onlineSavingsMsg'
Learn More

Cathepsin 6 Antibody (RM0157-3L27), Allophycocyanin, Novus Biologicals™

Product Code. 18404092 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18404092 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18404092 Supplier Novus Biologicals Supplier No. NBP212423APC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Cathepsin 6 Monoclonal antibody specifically detects Cathepsin 6 in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cathepsin 6
Applications Western Blot
Classification Monoclonal
Clone RM0157-3L27
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias 1600022N02Rik
Host Species Rat
Immunogen Mouse recombinant Cathepsin 6
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58518
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.