missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cathelicidin Recombinant Rabbit Monoclonal Antibody (K01_1N95)

Description
Cathelicidin Recombinant Monoclonal Antibody for Western Blot
Cathelicidin is an epithelial-derived antimicrobial protein found in specific granules of polymorphonuclear leukocytes of bone marrow, testis and neutrophils. Cathelicidins are one family of antimicrobial peptides characterized by conserved pro-peptide sequences that have been identified in epithelial tissues and some myeloid cells of humans and animals. LL-37/hCAP-18 is the only Cathelicidin found in humans and is expressed in inflammatory and epithelial cells. The presence of these molecules is essential for defense against invasive bacterial infection in skin. Besides their direct antimicrobial function, Cathelicidins have multiple roles in mediating innate and adaptive immunity, such as endotoxin neutralizing, angiogenesis, wound healing and promoting neutrophil chemotaxis and mast cell recruitment. Cathelicidin antimicrobial peptides qualify as prototypes of innovative drugs that may be used to treat infection and/or modulate the immune response.
Specifications
Specifications
| Antigen | Cathelicidin |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_1N95 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 40% glycerol, 150mM NaCl, 0.05% BSA and 0.01% sodium azide; pH 7.4 |
| Gene | CAMP |
| Gene Accession No. | P49913 |
| Gene Alias | 18 kDa cationic antimicrobial protein; Antibacterial peptide FALL-39; Antibacterial peptide LL-37; Camp; CAP18; CAP-18; Cathelicidin antimicrobial peptide; cathelin-like protein; Cathelin-related antimicrobial peptide; CLP; Cnlp; CRAMP; FALL39; FALL-39; FALL-39 peptide antibiotic; hCAP18; hCAP-18; HSD26; LL37; MCLP |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?