missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathelicidin Polyclonal Antibody, Invitrogen™
Description
Reconstitute with 500 μL of distilled water. OSC00028G can be used with blocking peptide GSC00002P.
Specifications
Specifications
| Antigen | Cathelicidin |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Antibody with no preservative |
| Gene | CAMP |
| Gene Accession No. | P49913 |
| Gene Alias | CAMP, CAP18, FALL39, FALL-39, 18 kDa cationic antimicrobial protein, CAP-18, hCAP-18, LL37, CRAMP, HSD26, hCAP18 |
| Gene Symbols | CAMP |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?