missing translation for 'onlineSavingsMsg'
Learn More

β-Catenin Mouse, Unlabeled, Clone: 14, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610154

 View more versions of this product

Product Code. 15825299

  • ONLINE EXCLUSIVE PRICE
    £311.00
    £307.25 / 1 set
    Save £3.75
    1% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

β-Catenin is a 92kDa protein that binds to the cytoplasmic tail of E-Cadherin. The cadherins, transmembrane adhesion molecules, are found with catenins at adherens junctions (zonula adherens). Deletions in the cytoplasmic domain of E-Cadherin which eliminate catenin binding also result in a loss of cell adhesion, indicating that this binding is essential for E-Cadherin function. Although the α- and β-Catenins have been cloned, very little is known about their biochemical roles. However a link between β-Catenin and colon cancer has been described. β-Catenin was found to co-immunoprecipitate with the APC tumor suppressor protein in human colorectal tumor cell lines, as well as in human kidney 293 cells. E-Cadherin, however, was not detectable in these complexes. Thus the APC-Catenin complex may be affecting the transmission of contact inhibition signals and/or the regulation of cell adhesion.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

β-Catenin
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Mouse β-Catenin aa. 571-781
150 μg
Cell Biology
Canine, Chicken, Human, Mouse, Rat
IgG1
Western Blot
14
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.