missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Catenin alpha-1 Recombinant Rabbit Monoclonal Antibody (9O1V3)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535193
This item is not returnable.
View return policy
Description
Immunogen sequence: MTAVHAGNIN FKWDPKSLEI RTLAVERLLE PLVTQVTTLV NTNSKGPSNK KRGRSKKAHV LAASVEQATE NFLEKGDKIA KESQFLKEEL VAAVEDVRKQ.
Associates with the cytoplasmic domain of a variety of cadherins. The association of catenins to cadherins produces a complex which is linked to the actin filament network, and which seems to be of primary importance for cadherins cell-adhesion properties. Can associate with both E- and N-cadherins. Originally believed to be a stable component of E-cadherin/catenin adhesion complexes and to mediate the linkage of cadherins to the actin cytoskeleton at adherens junctions. In contrast, cortical actin was found to be much more dynamic than E-cadherin/catenin complexes and CTNNA1 was shown not to bind to F-actin when assembled in the complex suggesting a different linkage between actin and adherence junctions components. The homodimeric form may regulate actin filament assembly and inhibit actin branching by competing with the Arp2/3 complex for binding to actin filaments. May play a crucial role in cell differentiation.
Specifications
| Catenin alpha-1 | |
| Recombinant Monoclonal | |
| 0.27 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P26231, P35221 | |
| CTNNA1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human alpha-Catenin (P35221). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 9O1V3 | |
| Unconjugated | |
| CTNNA1 | |
| [a]E-catenin; 102 kDa cadherin-associated protein; 2010010M04Rik; AA517462; AI988031; alpha E catenin; alpha E-catenin; alpha(E)-catenin; alpha-E-catenin; cadherin associated protein; Cadherin-associated protein; CAP102; catenin (cadherin associated protein), alpha 1; catenin (cadherin-associated protein), alpha; catenin (cadherin-associated protein), alpha 1; catenin (cadherin-associated protein), alpha 1, 102kDa; catenin alpha 1; catenin alpha 1 S homeolog; catenin alpha-1; catenin, alpha 1; Catna1; cell-adhesion protein alphaE catenin; ctnna; ctnna1; ctnna1.S; MDPT2; Renal carcinoma antigen NY-REN-13; XELAEV_18020018mg | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 12385, 1495, 307505 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction