missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Catechol Violet

Product Code. 11455517
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455517 5 g 5g
11465517 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11455517 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' B23513.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 115-41-3 | C19H14O7S | 386.37 g/mol

Catechol Violet is used as a complexometric indicator dye. It inhibits the synthesis of globin in reticulocyte lysates. It is also used as a pH indicator with a transition interval from 0 to 8.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Catechol Violet is used as a complexometric indicator dye. It inhibits the synthesis of globin in reticulocyte lysates. It is also used as a pH indicator with a transition interval from 0 to 8.

Solubility
Soluble in water, ethylene glycol, monomethyl ether and ethanol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-41-3
Molecular Formula C19H14O7S
Molecular Weight (g/mol) 386.37
MDL Number MFCD00005868
InChI Key RRRCKIRSVQAAAS-UHFFFAOYSA-N
Synonym Pyrocatechol sulfonephthalein; Pyrocatechol Violet
PubChem CID 66993
IUPAC Name 4-[3-(3,4-dihydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]benzene-1,2-diol
SMILES OC1=CC=C(C=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(O)=C1

Specifications

Beilstein 353938
Solubility Information Soluble in water,ethylene glycol,monomethyl ether and ethanol.
Formula Weight 386.38
Quantity 5 g
Chemical Name or Material Catechol Violet

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.