missing translation for 'onlineSavingsMsg'
Learn More

(-)-Catechin gallate, MedChemExpress

Product Code. 16418958
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16418958 10 mg 10mg
16411388 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16418958 Supplier Medchem Express Supplier No. HYN0356
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress (-)-Catechin gallate is a minor constituent in green tea catechins. (-)-Catechin gallate inhibits the activity of COX-1 and COX-2 enzymes.

(-)-Catechin gallate is a minor constituent in green tea catechins. (-)-Catechin gallate inhibits the activity of COX-1 and COX-2 enzymes.

Specifications

Chemical Name or Material (-)-Catechin gallate
CAS 130405-40-2
Color Pink
Purity Grade Notes Research
Molecular Formula C22H18O10
Quantity 10 mg
Synonym (-)-Catechin 3-gallate (-)-Catechin 3-O-gallate
Solubility Information DMSO : 100 mg/mL (226.06 mM; Need ultrasonic)
SMILES O=C(O[C@H]1[C@H](C2=CC=C(O)C(O)=C2)OC3=CC(O)=CC(O)=C3C1)C4=CC(O)=C(O)C(O)=C4
Molecular Weight (g/mol) 442.37
Formula Weight 442.37
Percent Purity 98.63%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.