Learn More
Fisherbrand™ Catalyst Norprene™ Food Formulation Pump Tubing
Available in standard 15m Lengths.
Brand: Fisherbrand™
Additional Details : Weight : 0.49000kg
Description
- Includes platinum-cured silicone, two TPEs and long-life PVC tubing material
- Available in both thin and thick wall configurations
Specifications
Catalyst pump tubing | |
3.2 mm | |
1.6 mm |