missing translation for 'onlineSavingsMsg'
Learn More

Catalpol, Thermo Scientific™

Product Code. 17565565 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity unitSize
17565565 250 mg 250mg
1 options
This item is not returnable. View return policy

Product Code. 17565565

Brand: Thermo Scientific Acros 468942500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 2415-24-9
Quantity 250 mg
Molecular Formula C15 H22 O10
Formula Weight 207
MDL Number MFCD11111524
Physical Form Powder
Color White to Light-brown
Purity ≥95%
InChI Key LHDWRKICQLTVDL-PZYDOOQISA-N
SMILES OC[C@H]1O[C@@H](O[C@@H]2OC=C[C@H]3[C@H](O)[C@@H]4O[C@]4(CO)[C@@H]23)[C@H](O)[C@@H](O)[C@@H]1O
IUPAC Name (2S,3R,4S,5S,6R)-2-{[(1S,2S,4S,5S,6R,10S)-5-hydroxy-2-(hydroxymethyl)-3,9-dioxatricyclo[4.4.0.0²,⁴]dec-7-en-10-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol) 362.33
Chemical Name or Material Catalpol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.