missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Catalase Recombinant Rabbit Monoclonal Antibody (24GB4295)

Rabbit Recombinant Monoclonal Antibody
£172.00 - £488.00
Specifications
| Antigen | Catalase |
|---|---|
| Clone | 24GB4295 |
| Concentration | 0.5 mg/mL |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275152
![]() |
Invitrogen™
MA557355 |
100 μL |
£488.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30274944
|
Invitrogen™
MA557356 |
20 μL |
£172.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Catalase Recombinant Monoclonal Antibody for Western Blot, Flow
Catalase is a homotetrameric heme-containing enzyme present within the matrix of all peroxisomes. It carries out a dismutation reaction in which hydrogen peroxide is converted to water and oxygen. Human catalase has the last four amino acids (-KANL) at the extreme C-terminus for peroxisome targeting. The monomer of human catalase is 61.3 kDa in molecular size. Catalase has been implicated as an important factor in inflammation, mutagenesis, prevention of apoptosis, and stimulation of a wide spectrum of tumors. Loss of catalase leads to the human genetic disease, acatalasemia, or Takahara's disease.Specifications
| Catalase | |
| 0.5 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P04040 | |
| 847 | |
| A synthesized peptide derived from human Catalase (aa 425-438). | |
| Antibody |
| 24GB4295 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| Cat | |
| 2210418N07; Cas1; Cas-1; CAT; Cat01; catalase; catalase 1; Catl; CS1; Cs-1; CTA1; HGNC:1516; hypothetical protein LOC531682; MGC138422; MGC138424 | |
| Cat | |
| Primary | |
| Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title