missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Catalase Rabbit anti-Human, Mouse, Rat, Clone: S03-8E9, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Catalase Monoclonal antibody specifically detects Catalase in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
Specifications
Specifications
| Antigen | Catalase |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Monoclonal |
| Clone | S03-8E9 |
| Conjugate | Unconjugated |
| Dilution | Western Blot, ELISA, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 2.5 μg/mL, Immunofluorescence 2.5 μg/mL |
| Formulation | 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide |
| Gene Alias | catalase, EC 1.11.1.6, MGC138422, MGC138424 |
| Host Species | Rabbit |
| Immunogen | Recombinant protein of mouse Catalase (Uniprot#: P24270) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?