missing translation for 'onlineSavingsMsg'
Learn More

Catalase Rabbit anti-Human, Clone: S05-5D9, Novus Biologicals™

Product Code. 18301912
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18301912 25 μg 25µL
18335212 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18301912 Supplier Bio-Techne Supplier No. NBP31491525UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Catalase Monoclonal antibody specifically detects Catalase in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Catalase
Applications Western Blot, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone S05-5D9
Conjugate Unconjugated
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
Gene Alias catalase, EC 1.11.1.6, MGC138422, MGC138424
Host Species Rabbit
Immunogen A synthetic peptide of human Catalase (Uniprot#: P04040)
Purification Method Affinity-purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Hypoxia, Lipid and Metabolism, Neuroscience, Peroxisome Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 847
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.