missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Catalase Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA586310

Product Code. 16393195

  • £449.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Purity is > 95% by SDS-PAGE.

Catalase is a homotetrameric heme-containing enzyme present within the matrix of all peroxisomes. It carries out a dismutation reaction in which hydrogen peroxide is converted to water and oxygen. Human catalase has the last four amino acids (-KANL) at the extreme C-terminus for peroxisome targeting. The monomer of human catalase is 61.3 kDa in molecular size. Catalase has been implicated as an important factor in inflammation, mutagenesis, prevention of apoptosis, and stimulation of a wide spectrum of tumors. Loss of catalase leads to the human genetic disease, acatalasemia, or Takahara's disease.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Catalase
Polyclonal
Unconjugated
Cat
2210418N07; Cas1; Cas-1; CAT; Cat01; catalase; catalase 1; Catl; CS1; Cs-1; CTA1; HGNC:1516; hypothetical protein LOC531682; MGC138422; MGC138424
Rabbit
Antigen Affinity Chromatography
RUO
12359, 24248, 847
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 0.05% sodium azide; pH 7.2
P04040, P04762, P24270
Cat
Synthetic peptide, corresponding to amino acids 341-390 of Human Catalase.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.