missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Catalase (Bovine)
Click to view available options
Quantity:
2 g
Unit Size:
2g
Description
#REF!
Specifications
Specifications
| Accession Number | P00432 |
| For Use With (Application) | Enzyme Activity |
| Gene ID (Entrez) | 531682 |
| Molecular Weight (g/mol) | 240kDa |
| Name | Catalase (Bovine) |
| Preparation Method | Native protein purified from Bovine Liver |
| Quantity | 2 g |
| Storage Requirements | Store at -20°C on dry atmosphere. |
| Regulatory Status | RUO |
| Gene Alias | MGC128112 |
| Show More |
Safety and Handling
Product Identifier
- 2GR Catalase
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P233-Keep container tightly closed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P404-Store in a closed container.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : sodium phosphate monobasic, potassium phosphate dibasic
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction