missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Catalase Antibody (HPYL/7171) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14082JF669
This item is not returnable.
View return policy
Description
Catalase Monoclonal antibody specifically detects Catalase in H. pylori samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Catalase | |
| Monoclonal | |
| Janelia Fluor 669 | |
| catalase, EC 1.11.1.6, MGC138422, MGC138424 | |
| Recombinant Helicobacter pylori Catalase protein fragment (around aa323-445) (Exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer, Cellular Markers, Hypoxia, Lipid and Metabolism, Neuroscience, Peroxisome Markers, Signal Transduction | |
| 847 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| HPYL/7171 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| H. pylori | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction