missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-59857
This item is not returnable.
View return policy
Description
CAT1 Polyclonal specifically detects CAT1 in Human samples. It is validated for Western Blot.
Specifications
| CAT1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| amino acid transporter, cationic 1, ATRC1ERR, CAT-1System Y+ basic amino acid transporter, Ecotropic retroviral leukemia receptor homolog, ecotropic retroviral receptor, Ecotropic retrovirus receptor homolog, HCAT1, high affinity cationic amino acid transporter 1, REC1LCAT1, solute carrier family 7 (cationic amino acid transporter, y+ system), member 1, Solute carrier family 7 member 1 | |
| Rabbit | |
| 68 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 100%; Equine: 92%; Pig: 85%. | |
| Human, Pig, Bovine, Equine, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| P30825 | |
| SLC7A1 | |
| The immunogen for anti-SLC7A1 antibody: synthetic peptide directed towards the N terminal of human SLC7A1. Peptide sequence LGFIMVSGFVKGSVKNWQLTEEDFGNTSGRLCLNNDTKEGKPGVGGFMPF The peptide sequence for this immunogen was taken from within the described region. | |
| Affinity purified | |
| RUO | |
| 6541 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto