missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAT1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32271AF647
This item is not returnable.
View return policy
Description
CAT1 Polyclonal antibody specifically detects CAT1 in Human samples. It is validated for Western Blot
Specifications
| CAT1 | |
| Polyclonal | |
| Western Blot | |
| amino acid transporter, cationic 1, ATRC1ERR, CAT-1System Y+ basic amino acid transporter, Ecotropic retroviral leukemia receptor homolog, ecotropic retroviral receptor, Ecotropic retrovirus receptor homolog, HCAT1, high affinity cationic amino acid transporter 1, REC1LCAT1, solute carrier family 7 (cationic amino acid transporter, y+ system), member 1, Solute carrier family 7 member 1 | |
| The immunogen this antibody was made to, maps to a region between residue 600 to 650 of human High Affinity Cationic Amino Acid Transporter 1 using the numbering given in entry NP_003036.1 (GeneID 6541). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 6541 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction