missing translation for 'onlineSavingsMsg'
Learn More

CASZ1 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18326014 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18326014 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18326014 Supplier Novus Biologicals Supplier No. NBP306281AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CASZ1 Polyclonal specifically detects CASZ1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CASZ1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias castor, castor homolog 1, zinc finger, castor homolog 1, zinc finger (Drosophila), castor zinc finger 1, Castor-related protein, CST, dJ734G22.1, FLJ20321, Putative survival-related protein, SRG, SRGcst, survival related, survival-related, Zinc finger protein 693, zinc finger protein castor homolog 1, ZNF693, ZNF693FLJ12223
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CASZ1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54897
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.