missing translation for 'onlineSavingsMsg'
Learn More

Castor Oil, Technical Grade, TRC

Product Code. 30355924
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
Product Code. Quantity unitSize
30355924 25 mL 25mL
30350492 100 mL 100mL
2 options
This item is not returnable. View return policy

Product Code. 30355924

Brand: Toronto Research Chemicals TRCC18480525ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 8001-79-4
InChI Formula InChI=1S/C57H104O9/c1-4-7-10-31-40-51(58)43-34-25-19-13-16-22-28-37-46-55(61)64-49-54(66-57(63)48-39-30-24-18-15-21-27-36-45-53(60)42-33-12-9-6-3)50-65-56(62)47-38-29-23-17-14-20-26-35-44-52(59)41-32-11-8-5-2/h25-27,34-36,51-54,58-60H,4-24,28-33,37-50H2,1-3H3/b34-25-,35-26-,36-27-
IUPAC Name propane-1,2,3-triyl (9Z,9'Z,9''Z)-tris(12-hydroxyoctadec-9-enoate)
SMILES CCCCCCC(O)C/C=C\CCCCCCCC(=O)OCC(OC(=O)CCCCCCC/C=C\CC(O)CCCCCC)COC(=O)CCCCCCC/C=C\CC(O)CCCCCC
Synonym 3 Castor Oil, AS 15, Acrol 1070, Aromatic castor oil, Recastol 2790, Ricinus communis oil, Thixcin ST, Viscotrol C, Neoloid, LAV (polyol), Thixcin ST, Ricinus communis oil, Oil of Palma Christi
Chemical Name or Material Castor oil

Specifikationer

Quantity 25 mL
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.