missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Castor oil, specified according to the requirements of Ph.Eur.

Product Code. 10566051 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
2.5 L
500mL
This item is not returnable. View return policy

Product Code. 10566051

Brand: Thermo Scientific Chemicals 348855000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 8001-79-4 | C57H104O9 | 933.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 8001-79-4
Molecular Formula C57H104O9
Molecular Weight (g/mol) 933.45
MDL Number MFCD00130746
InChI Key ZEMPKEQAKRGZGQ-AAKVHIHISA-N
Synonym castor oil, ricinus oil, olio di ricino, venelex, xenaderm, optase, trypsin complex, unii-d5340y2i9g, castor oil usp:jan, 1-o,2-o,3-o-tris z-12-hydroxy-9-octadecenoyl glycerol
PubChem CID 14030006
IUPAC Name 2,3-bis[[(Z)-12-hydroxyoctadec-9-enoyl]oxy]propyl (Z)-12-hydroxyoctadec-9-enoate
SMILES CCCCCCC(CC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCC(CCCCCC)O)OC(=O)CCCCCCCC=CCC(CCCCCC)O)O

Specifikationer

Acidity 2.0mg KOH/g max.
Absorbance 1.5 max. (at the absorption maximum at 269 +/- 1nm)
Melting Point -10.0°C
Color Colorless or Yellow
Density 0.9570g/mL
Boiling Point 229.0°C
Flash Point 229°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4770 to 1.4810
Quantity 500 mL
Merck Index 14,1898
Specific Gravity 0.957
Specific Rotation + 3.50
Solubility Information Solubility in water: .. Other solubilities: miscible with alcohol, benzene, chloroform,, carbondisulfide, ether and glacial acetic acid
Viscosity 0,6-0,8 Pas (25°C)
Grade Eu. Ph.
Concentration or Composition (by Analyte or Components) of fatty acids (GC): Passes Test (per Eur. Pharm.)
Physical Form Viscous Hygroscopic Liquid
Chemical Name or Material Castor oil
Visa mer Visa mindre

RUO – Research Use Only

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.