missing translation for 'onlineSavingsMsg'
Learn More

Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals

£19.00 - £119.00

Chemical Identifiers

CAS 8001-79-4
Molecular Formula C57H104O9
Molecular Weight (g/mol) 933.45
MDL Number MFCD00130879
InChI Key ZEMPKEQAKRGZGQ-UHFFFAOYNA-N
IUPAC Name 1,3-bis[(12-hydroxyoctadec-9-enoyl)oxy]propan-2-yl 12-hydroxyoctadec-9-enoate
SMILES CCCCCCC(O)CC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCC(O)CCCCCC)OC(=O)CCCCCCCC=CCC(O)CCCCCC
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10295234
View Documents Promotion Details
Thermo Scientific Acros
397282500
250 g
£42.70
250g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10197170
View Documents Promotion Details
Thermo Scientific Acros
404165000
500 g
£19.00
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10422514
View Documents Promotion Details
Thermo Scientific Acros
397280010
1 kg
£119.00
1kg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10539813
View Documents Promotion Details
Thermo Scientific Acros
404160025
2.5 kg
£55.00
2.50kg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

8001-79-4
933.45
ZEMPKEQAKRGZGQ-UHFFFAOYNA-N
CCCCCCC(O)CC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCC(O)CCCCCC)OC(=O)CCCCCCCC=CCC(O)CCCCCC
C57H104O9
MFCD00130879
1,3-bis[(12-hydroxyoctadec-9-enoyl)oxy]propan-2-yl 12-hydroxyoctadec-9-enoate

Specifications

8001-79-4
Yellow
>100.0°C
Authentic
C57H104O9
176 to 182g KOH/g
Solubility in water: 100g/L (20°C). Other solubilities: soluble in organic solvents
CCCCCCC(O)CC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCC(O)CCCCCC)OC(=O)CCCCCCCC=CCC(O)CCCCCC
933.45
Viscous Liquid
4.0°C
1.0500g/mL
257°C
Glass bottle
MFCD00130879
1.05
ZEMPKEQAKRGZGQ-UHFFFAOYNA-N
1,3-bis[(12-hydroxyoctadec-9-enoyl)oxy]propan-2-yl 12-hydroxyoctadec-9-enoate
650-800 mPa.s (25°C)
Castor oil
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 500-151-7

missing translation for 'rtecsNumber' : FI4100000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only