missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CASRegistryNumber1t Iron, Chromium, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
CASRegistryNumber1t Iron, Chromium, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Lead/Sulfur/Tin/Phosphorus/Molybdenum/Copper/Cobalt/Vanadium/Tungsten/Nickel/Silicon/Manganese/Carbon/Chromium/Aluminium |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Lead 0.012 %/Sulfur 0.0297 %/Tin 0.036 %/Phosphorus 0.082 %/Molybdenum 0.091 %/Copper 0.103 %/Cobalt 0.128 %/Vanadium 0.132 %/Tungsten 0.265 %/Nickel 0.309 %/Silicon 0.365 %/Manganese 0.451 %/Carbon 2.76 %/Chromium 29.31 %/Aluminium (0.147) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction