missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CASS4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568383
This item is not returnable.
View return policy
Description
CASS4 Polyclonal Antibody for IHC (P)
Docking protein that plays a role in tyrosine kinase-based signaling related to cell adhesion and cell spreading. Regulates PTK2/FAK1 activity, focal adhesion integrity, and cell spreading. [UniProt].
Specifications
| CASS4 | |
| Polyclonal | |
| Unconjugated | |
| CASS4 | |
| C20orf32; cas scaffolding protein family member 4; CAS4; CASS4; HEF1-EFS-p130Cas-like protein; HEFL; HEF-like protein; HEPL | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57091 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.9 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9NQ75 | |
| CASS4 | |
| Synthetic peptide corresponding to residues near the C terminal of human CASS4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction