missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CASRegistryNumber1t Iron, Low Alloy, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
CASRegistryNumber1t Iron, Low Alloy, Ø38x15mm
Specifications
Specifications
| Chemical Name or Material | Lead/Nitrogen/Aluminium/Arsenic/Antimony/Tin/Titanium/Cobalt/Phosphorus/Vanadium/Molybdenum/Tungsten/Chromium/Copper/Manganese/Nickel/Silicon/Carbon |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Lead 0.003 %/Nitrogen 0.006 %/Aluminium 0.02 %/Arsenic 0.022 %/Antimony 0.035 %/Tin 0.037 %/Titanium 0.045 %/Cobalt 0.06 %/Phosphorus 0.1 %/Vanadium 0.1 %/Molybdenum 0.29 %/Tungsten 0.32 %/Chromium 0.42 %/Copper 0.63 %/Manganese 0.75 %/Nickel 0.85 %/Silicon 1.85 %/Carbon 3.35 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction