missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspr2 Antibody (840727), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5145M
This item is not returnable.
View return policy
Description
Caspr2 Monoclonal antibody specifically detects Caspr2 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Caspr2 | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 840727 | |
| Flow Cytometry, CyTOF-ready | |
| AUTS15, CASPR2DKFZp781D1846, CDFE, Cell recognition molecule Caspr2, contactin associated protein-like 2, contactin-associated protein 2, contactin-associated protein-like 2, homolog of Drosophila neurexin IV, KIAA0868, NRXN4Caspr2, PTHSL1 | |
| Mouse myeloma cell line NS0-derived recombinant human Caspr2, Pro29-Ala1262, Accession # Q9UHC6 | |
| 0.1 mL | |
| Neuroscience | |
| 26047 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction