missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Caspr2 Monoclonal antibody specifically detects Caspr2 in Mouse samples. It is validated for ELISA
Specifications
Specifications
| Antigen | Caspr2 |
| Applications | ELISA |
| Clone | 001 |
| Conjugate | Janelia Fluor 549 |
| Gene Alias | AUTS15, CASPR2DKFZp781D1846, CDFE, Cell recognition molecule Caspr2, contactin associated protein-like 2, contactin-associated protein 2, contactin-associated protein-like 2, homolog of Drosophila neurexin IV, KIAA0868, NRXN4Caspr2, PTHSL1 |
| Gene ID (Entrez) | 26047 |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?