missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspr1 Rabbit anti-Human, Mouse, Rat, Clone: 8O5H6, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | Caspr1 |
|---|---|
| Clone | 8O5H6 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18398282
|
Novus Biologicals
NBP3-16262-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18383203
|
Novus Biologicals
NBP3-16262-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Caspr1 Monoclonal antibody specifically detects Caspr1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Caspr1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 8506 | |
| IgG | |
| Affinity purified |
| 8O5H6 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Caspr, Caspr1, CASPRcaspr1, CNTNAP, contactin associated protein 1, neurexin 4, Neurexin IV, Neurexin-4, NRXN4contactin-associated protein 1, P190 | |
| A synthetic peptide corresponding to a sequence within amino acids 700-800 of human Caspr11 (P78357). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title