missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase 9/p35/p10 Mouse anti-Human, Mouse, Clone: 1B7G2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66169-1-IG-20UL
This item is not returnable.
View return policy
Description
Involved in the activation cascade of caspases responsible for apoptosis execution. Binding of caspase-9 to Apaf-1 leads to activation of the protease which then cleaves and activates caspase-3. Promotes DNA damage-induced apoptosis in a ABL1/c-Abl-dependent manner. Proteolytically cleaves poly(ADP-ribose) polymerase (PARP).Specifications
| Caspase 9/p35/p10 | |
| Monoclonal | |
| 1.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P55211, Q8C3Q9 | |
| CASP9 | |
| Caspase 9/p35/p10 Fusion Protein Ag20813 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG2b |
| Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1B7G2 | |
| Unconjugated | |
| CASP9 | |
| APAF 3, APAF3, Apoptotic protease Mch 6, CASP 9, CASP9, Caspase 9, Caspase 9/p35/p10, CASPASE 9c, Caspase9, Caspase-9, ICE LAP6, ICE like apoptotic protease 6, MCH6 | |
| Mouse | |
| Protein A | |
| RUO | |
| 12371, 842 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction