missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Caspase 9 (Cleaved Asp315) Recombinant Rabbit Monoclonal Antibody (23GB725), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | Caspase 9 (Cleaved Asp315) |
|---|---|
| Clone | 23GB725 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089952
![]() |
Invitrogen™
MA553678 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088375
|
Invitrogen™
MA553679 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Caspase 9 (Cleaved Asp315) Recombinant Monoclonal antibody specifically detects Caspase 9 (Cleaved Asp315) in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| Caspase 9 (Cleaved Asp315) | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P55211, Q8C3Q9 | |
| 12371, 842 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB725 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| APAF3; APAF-3; apoptosis-related cysteine protease; apoptotic protease activating factor 3; apoptotic protease MCH-6; Apoptotic protease-activating factor 3; cas9; CASP9; CASP-9; caspase 9; caspase 9, apoptosis-related cysteine peptidase; caspase-9; Caspase-9 subunit p10; Caspase-9 subunit p35; ICE-LAP6; ICE-like apoptotic protease 6; MCH6; OTTHUMP00000002324; PPP1R56; protein phosphatase 1, regulatory subunit 56 | |
| A synthetic peptide of human CASP9 (90-350AA). | |
| Primary | |
| -20°C | |
| CASP9 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title