missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-9 Antibody (LAP6 96-2-22) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB500-209JF669
This item is not returnable.
View return policy
Description
Caspase-9 Monoclonal antibody specifically detects Caspase-9 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Simple Western,KnockDown,Immunocytochemistry/ Immunofluorescence
Specifications
| Caspase-9 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| APAF-3APAF3, apoptotic protease activating factor 3, Apoptotic protease Mch-6, Apoptotic protease-activating factor 3, CASP-9, caspase 9, apoptosis-related cysteine peptidase, caspase-9, EC 3.4.22.62, ICE-LAP6CASPASE-9c, ICE-like apoptotic protease 6, MCH6apoptosis-related cysteine protease | |
| Recombinant human Caspase 9 pro-domain protein. [UniProt# P55211] | |
| 0.2 mL | |
| Alzheimers Research, Apoptosis, Cancer, Caspases, Cell Biology, Neurodegeneration, Neuroscience, Phospho Specific | |
| 842 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Simple Western, KnockDown, Immunocytochemistry/Immunofluorescence | |
| LAP6 96-2-22 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction