missing translation for 'onlineSavingsMsg'
Learn More

Caspase-9 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18401854 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18401854 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18401854 Supplier Novus Biologicals Supplier No. NBP176961AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Caspase-9 Polyclonal antibody specifically detects Caspase-9 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase-9
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias APAF-3APAF3, apoptotic protease activating factor 3, Apoptotic protease Mch-6, Apoptotic protease-activating factor 3, CASP-9, caspase 9, apoptosis-related cysteine peptidase, caspase-9, EC 3.4.22.62, ICE-LAP6CASPASE-9c, ICE-like apoptotic protease 6, MCH6apoptosis-related cysteine protease
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 20 amino acids near the center of human Caspase 9. The immunogen is located within amino acids 290-340 of Caspase 9. Amino Acid Sequence: ASTSPEDESPGSNPEPDATP
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cancer, Caspases, Cell Biology, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 842
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.