missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Caspase 8 (Cleaved Asp374) Recombinant Rabbit Monoclonal Antibody (RM442)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA538680
This item is not returnable.
View return policy
Description
This product targets amino acids 375-479 of Caspase 8.
Caspase 8 (CASP8) is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases play a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes composed of a pro-domain, a large protease subunit, and a small protease subunit. Activation of caspases requires proteolytic processing at conserved internal aspartic residues to generate a heterodimeric enzyme consisting of the large and small subunits. Caspase 8 is involved in the programmed cell death induced by Fas and various apoptotic stimuli. The N-terminal FADD-like death effector domain of Caspase 8 suggests that it may interact with Fas-interacting protein FADD. Caspase 8 was detected in the insoluble fraction of the affected brain region from Huntington disease patients but not in those from normal controls, which implicated the role in neurodegenerative diseases. Caspase 8 binds to the death effector domain (DED) of FADD through an analogous DED domain present in tandem in the pro-form of the Caspase 8 protein. Activated Caspase 8 then activates other downstream caspases including Caspase 9, thereby committing the cell to undergo apoptosis. In addition, Caspase 8 also reacts with Jurkat cells and Tonsil. Overexpression of Caspase 8 induces apoptosis, which can be blocked by inhibitors specific for the ICE family. Many alternatively spliced transcript variants encoding different isoforms have been described for Caspase 8, however, not all variants have had their full-length sequences determined.
Specifications
| Caspase 8 (Cleaved Asp374) | |
| Recombinant Monoclonal | |
| 50 μg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide | |
| Q14790 | |
| CASP8 | |
| A peptide corresponding to the C-terminous of caspase 8. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| RM442 | |
| Unconjugated | |
| CASP8 | |
| ALPS2B; Apoptotic cysteine protease; apoptotic protease Mch-5; CAP4; CASP8; Casp-8; caspase 8; caspase 8, apoptosis-related cysteine peptidase; caspase 8, apoptosis-related cysteine protease; caspase-8; Caspase-8 subunit p10; Caspase-8 subunit p18; FADD-homologous ICE/ced-3-like protease; FADD-like ICE; FLICE; ICE-like apoptotic protease 5; MACH; MACH-alpha-1/2/3 protein; MACH-beta-1/2/3/4 protein; MCH5; MORT1-associated ced-3 homolog | |
| Rabbit | |
| Protein A | |
| RUO | |
| 841 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction