missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase 7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27155-1-AP-150UL
This item is not returnable.
View return policy
Description
Caspases are a family of cysteine proteases that can be divided into the apoptotic and inflammatory caspase subfamilies. Unlike the apoptotic caspases, members of the inflammatory subfamily are generally not involved in cell death but are associated with the immune response to microbial pathogens. The apoptotic subfamily can be further divided into initiator caspases, which are activated in response to death signals, and executioner caspases, which are activated by the initiator caspases and are responsible for cleavage of cellular substrates that ultimately lead to cell death. Caspase-7 is an executioner caspase that was identified based on its homology with caspases 1 and 3, as well as the C. elegans cell death protein CED-3. Alternative splicing of Caspase-7 mRNA results in the production of 3 distinct isoforms. Caspase-7 activity can be directly inhibited by XIAP expression.Specifications
| Caspase 7 | |
| Polyclonal | |
| Unconjugated | |
| CASP7 | |
| Apoptotic protease Mch 3, CASP 7, CASP7, Caspase 7, Caspase7, CMH 1, ICE LAP3, ICE like apoptotic protease 3, MCH3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12369, 840 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P55210, P97864 | |
| CASP7 | |
| Caspase 7 Fusion Protein Ag25904 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction