missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-7 Rabbit anti-Human, Clone: S03-0F2, Novus Biologicals™
Description
Caspase-7 Monoclonal antibody specifically detects Caspase-7 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Caspase-7 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | S03-0F2 |
| Conjugate | Unconjugated |
| Dilution | Western Blot, ELISA, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunohistochemistry-Paraffin |
| Formulation | 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA |
| Gene Alias | apoptosis-related cysteine protease, Apoptotic protease Mch-3, CASP-7, caspase 7, apoptosis-related cysteine peptidase, caspase-7, EC 3.4.22, EC 3.4.22.60, ICE-like apoptotic protease 3, Lice2 alpha/beta/gamma |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide of human Caspase-7 (Uniprot#: P55210) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?