missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Caspase-7 Antibody (MCH3-5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-30814-100ug
This item is not returnable.
View return policy
Description
Caspase-7 Monoclonal antibody specifically detects Caspase-7 in Human samples. It is validated for Western Blot
Specifications
| Caspase-7 | |
| Monoclonal | |
| Unconjugated | |
| apoptosis-related cysteine protease, Apoptotic protease Mch-3, CASP-7, caspase 7, apoptosis-related cysteine peptidase, caspase-7, EC 3.4.22, EC 3.4.22.60, ICE-like apoptotic protease 3, Lice2 alpha/beta/gamma | |
| Hybridoma produced by the fusion of splenocytes from mice immunized with recombinant human Caspase-7 protein and mouse myeloma cells. | |
| 100 μg | |
| Apoptosis, Cancer, Caspases, Cell Biology, Cellular Markers, Ubiquitin Proteasome Pathway | |
| 840 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| MCH3-5 | |
| Phosphate buffered saline | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction