missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Caspase-7 Antibody (7-1-11), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-19229AF750
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Caspase-7 Monoclonal specifically detects Caspase-7 in Human, Mouse, Rat, Porcine, Canine, Hamster, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry.
Tekniske data
| Caspase-7 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CASP7 | |
| Recombinant human Caspase-7 protein (full length) | |
| 0.1 mL | |
| Apoptosis, Cancer, Caspases, Cell Biology, Cellular Markers, Ubiquitin Proteasome Pathway | |
| Caspase-7 (7-1-11) | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| 7-1-11 | |
| Western Blot, Immunohistochemistry | |
| apoptosis-related cysteine protease, Apoptotic protease Mch-3, CASP-7, caspase 7, apoptosis-related cysteine peptidase, caspase-7, EC 3.4.22, EC 3.4.22.60, ICE-like apoptotic protease 3, Lice2 alpha/beta/gamma | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Canine, Hamster, Primate, Rabbit, Sheep | |
| IgG2b |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion