missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Caspase 6 p18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578922
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Jurkat whole cell. IHC: human colorectal adenocarcinoma tissue, human pancreatic cancer tissue. Flow: SiHa cell.
Caspase 6 is an enzyme that in humans is encoded by the caspase 6 gene. This gene encodes a protein that is a member of the cysteine-aspartic acid protease (caspase) family. Using radiation hybrid mapping, the Caspase 6 gene is localized to human chromosome 4q25-q26. Caspase 6 functions as a downstream enzyme in the caspase activation cascade. And Caspase 6 can cleave lamin A to its signature apoptotic fragment.
Specifications
| Caspase 6 p18 | |
| Polyclonal | |
| Unconjugated | |
| CASP6 | |
| apoptotic protease MCH-2; CASP6; CASP-6; caspase 6; caspase 6, apoptosis-related cysteine peptidase; caspase 6, apoptosis-related cysteine protease; caspase-6; Caspase-6 subunit p11; Caspase-6 subunit p18; MCH2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 839 | |
| -20°C | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| P55212 | |
| CASP6 | |
| A synthetic peptide corresponding to a sequence at the N-terminus of human Caspase-6 (P18) (24-44aa AFYKREMFDPAEKYKMDHRRR). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction