Learn More
Invitrogen™ Caspase 6 Monoclonal Antibody (MCH2 14 1-190)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA112558
Product Code. 11588153
  • £440.00 / 100µg
Description
A suggested positive control for this product is MCF-7 cell lysate.
Caspases are cysteine proteases, expressed as inactive precursors, that mediate apoptosis by proteolysis of specific substrates. Caspases have the ability to cleave after aspartic acid residues. There are two classes of caspases involved in apoptosis; initiators (activation by receptor cluster) and effectors (activation by mitochondrial permeability transition). Proapoptotic signals autocatalytically activate initiator caspases, such as Caspase-8 and Caspase 9. Activated initiator caspases then process effector caspases, such as Caspase-3 and Caspase-7, which in turn cause cell collapse.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Caspase 6
Monoclonal
1 mg/mL
PBS with 0.08% sodium azide
P55212
Casp6
Recombinant human capase-6 protein.
100 μg
Primary
Human
Antibody
IgG
Western Blot
MCH2 14 1-190
Unconjugated
Casp6
apoptotic protease MCH-2; CASP6; CASP-6; caspase 6; caspase 6, apoptosis-related cysteine peptidase; caspase 6, apoptosis-related cysteine protease; caspase-6; Caspase-6 subunit p11; Caspase-6 subunit p18; mCASP-6; MCH2
Mouse
Protein A/G
RUO
839
-20°C, Avoid Freeze/Thaw Cycles