missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-6 Antibody (MCH2 14 1-190), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-66612
This item is not returnable.
View return policy
Description
Caspase-6 Monoclonal specifically detects Caspase-6 in Human samples. It is validated for Western Blot.
Specifications
| Caspase-6 | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.08% Sodium Azide | |
| Apoptotic protease Mch-2, CASP-6, caspase 6, apoptosis-related cysteine peptidase, caspase 6, apoptosis-related cysteine protease, caspase-6, EC 3.4.22, MCH2EC 3.4.22.59 | |
| Mouse | |
| Protein A or G purified | |
| Apoptosis, Cancer, Caspases, Phospho Specific | |
| 839 | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| MCH2 14 1-190 | |
| Western Blot 1:100-1:2000 | |
| P55212 | |
| CASP6 | |
| Hybridoma produced by the fusion of splenocytes from mice immunized with recombinant human capase-6 protein and mouse myeloma cells. | |
| 100 μg | |
| Primary | |
| Detects proenzyme form of human Caspase-6 by Western blot. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction