missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-6 Antibody (M378), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23183
This item is not returnable.
View return policy
Description
Caspase-6 Monoclonal antibody specifically detects Caspase-6 in Human samples. It is validated for Western Blot
Specifications
| Caspase-6 | |
| Monoclonal | |
| Unconjugated | |
| PBS, 1 mg/ml BSA and 50% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| M378 | |
| Western Blot 1:250 | |
| Apoptotic protease Mch-2, CASP-6, caspase 6, apoptosis-related cysteine peptidase, caspase 6, apoptosis-related cysteine protease, caspase-6, EC 3.4.22, MCH2EC 3.4.22.59 | |
| Clone (M378) was generated from a peptide containing amino acid residues in the C-terminal region of human active/cleaved Caspase-6. [UniProt# P55212] | |
| 0.1 mL | |
| Apoptosis, Cancer, Caspases, Phospho Specific | |
| 839 | |
| Store at -20°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction