missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-6 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99463AF532
This item is not returnable.
View return policy
Description
Caspase-6 Polyclonal antibody specifically detects Caspase-6 in Human samples. It is validated for Western Blot
Specifications
| Caspase-6 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 532 | |
| Apoptotic protease Mch-2, CASP-6, caspase 6, apoptosis-related cysteine peptidase, caspase 6, apoptosis-related cysteine protease, caspase-6, EC 3.4.22, MCH2EC 3.4.22.59 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Caspase-6. | |
| 0.1 mL | |
| Apoptosis, Cancer, Caspases | |
| 839 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction