missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Caspase 5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111126
This item is not returnable.
View return policy
Description
Immunogen sequence: VEGVFIFLIE DSGKKKRRKN FEAMFKGILQ SGLDNFVINH MLKNNVAGQT SIQTLVPNTD QKSTSVKKDN HKKKTVKMLE YLGKDV Highest antigen sequence indentity to the following orthologs: Mouse - 23%, Rat - 23%.
Caspases are a family of cysteine proteases that can be divided into the apoptotic and inflammatory caspase subfamilies. Unlike the apoptotic caspases, members of the inflammatory subfamily are generally not involved in cell death but are associated with the immune response to microbial pathogens. Members of this subfamily include caspase-1, -4, -5, and -12. Activation of these caspases results in the cleavage and activation of proinflammatory cytokines such as IL-1-b and IL-18. Caspase-5 can interact with caspase-1; both are constituents of the NALP1 inflammasome, a complex that can trigger the cleavage of pro-IL-1-b. Expression of caspase-5 can be regulated by lipopolysaccharide (LPS) and IFN-gamma.
Specifications
| Caspase 5 | |
| Polyclonal | |
| Unconjugated | |
| CASP5 | |
| CASP5; CASP-5; caspase 5; caspase 5, apoptosis-related cysteine peptidase; caspase 5, apoptosis-related cysteine protease; caspase-5; Caspase-5 subunit p10; Caspase-5 subunit p20; ICE(rel)III; ICE(rel)-III; ICEREL-III; ICH3; ICH-3; Protease ICH-3; protease TY; TY protease | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 838 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P51878 | |
| CASP5 | |
| Recombinant Protein corresonding to Human Caspase 5. Recombinant protein control fragment (Product #RP-98453). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction