missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase 5 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77209G
This item is not returnable.
View return policy
Description
Caspase 5 Polyclonal antibody specifically detects Caspase 5 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Specifications
| Caspase 5 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
| DyLight 488 | |
| CASP-5, caspase 5, apoptosis-related cysteine peptidase, caspase 5, apoptosis-related cysteine protease, caspase-5, EC 3.4.22, EC 3.4.22.58, ICE(rel)III, ICE(rel)-III, ICEREL-III, ICH3, ICH-3, MGC141966, Protease ICH-3, Protease TY, TY protease | |
| Antibody was raised against a 14 amino acid synthetic peptide from the center of human Caspase-5. The immunogen is located within amino acids 170 - 220 of Caspase-5. Amino Acid Squence: YPIKKREDRRRLAL | |
| 0.1 mL | |
| Apoptosis, Cancer, Caspases | |
| 838 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction