missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase 5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
£141.00
Specifications
| Antigen | Caspase 5 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 2 μg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
Description
Caspase 5 Polyclonal specifically detects Caspase 5 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| Caspase 5 | |
| Western Blot 0.5-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 2 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cancer, Caspases | |
| PBS with 0.02% Sodium Azide | |
| CASP-5, caspase 5, apoptosis-related cysteine peptidase, caspase 5, apoptosis-related cysteine protease, caspase-5, EC 3.4.22, EC 3.4.22.58, ICE(rel)III, ICE(rel)-III, ICEREL-III, ICH3, ICH-3, MGC141966, Protease ICH-3, Protease TY, TY protease | |
| CASP5 | |
| IgG | |
| Affinity Purified | |
| Depending on cell lines or tissues used, other cleavage products may be observed. |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| P51878 | |
| 838 | |
| Antibody was raised against a 14 amino acid synthetic peptide from the center of human Caspase-5. The immunogen is located within amino acids 170 - 220 of Caspase-5. Amino Acid Squence: YPIKKREDRRRLAL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title