missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase 4/p20/p10 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11856-1-AP-150UL
This item is not returnable.
View return policy
Description
Caspases are cysteine proteases, expressed as inactive precursors, that mediate apoptosis by proteolysis of specific substrates. Caspases have the ability to cleave after aspartic acid residues. There are two classes of caspases involved in apoptosis; initiators (activation by receptor cluster) and effectors (activation by mitochondrial permeability transition). Proapoptotic signals autocatalytically activate initiator caspases, such as Caspase-8 and Caspase 9. Activated initiator caspases then process effector caspases, such as Caspase-3 and Caspase-7, which in turn cause cell collapse.Specifications
| Caspase 4/p20/p10 | |
| Polyclonal | |
| Unconjugated | |
| CASP4 | |
| CASP 4, CASP4, Caspase 4, Caspase4, ICE(rel) II, ICE(rel)II, ICEREL II, ICH 2, ICH2, Mih1/TX, Protease ICH 2, Protease TX, TX | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 837 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P49662 | |
| CASP4 | |
| Caspase 4/p20/p10 Fusion Protein Ag2429 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction